missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl ether, ACS Reagent, contains either BHT or hydroquinone as stabilizer, ≥99.0%, Honeywell Riedel-de Haën™

Product Code. 15635380
Click to view available options
Quantity:
500 mL
6 x 1 L
Packaging:
Fiberboard box
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent, contains either BHT or hydroquinone as stabilizer,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Melting Point -86°C
Density 0.725g/mL at 25°C
Boiling Point 67°C to 70°C (1013 hPa)
Flash Point −29°C (−20.2°F)
Odor Odorless
Packaging Fiberboard box
Linear Formula (CH3)2CHOCH(CH3)2
Quantity 6 x 1 L
UN Number UN1159
Beilstein 1731256
Vapor Density 3.5 (vs air)
Formula Weight 102.17g/mol
Percent Purity ≥99%
Chemical Name or Material Diisopropyl ether
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'