missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (-)-Diisopropyl-L-malate, 97%

Product Code. 11455067
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 83541-68-8
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.23
MDL Number MFCD00274300
InChI Key DQOODBSVDOAUMP-JTQLQIEISA-L
Synonym diisopropyl s---malate, --diisopropyl-l-malate, 1,4-diisopropyl 2s-2-hydroxybutanedioate, diisopropyl d-malate, s-diisopropyl malate, diisopropyl-s---malate, l-malic acid diisopropyl ester, s-2-hydroxybutanedioic acid diisopropyl ester
PubChem CID 11333435
SMILES CC(C)O[C@@](CC([O-])=O)(C(C)C)C([O-])=O

Specifications

Density 1.055
Boiling Point 236°C to 238°C
Quantity 25 g
IUPAC Name (2S)-2-(propan-2-yl)-2-(propan-2-yloxy)butanedioate
Formula Weight 218.25
Percent Purity 97%
Chemical Name or Material (-)-Diisopropyl-L-malate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'