missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl phosphite, 98%, Thermo Scientific Chemicals

Product Code. 11470258
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Diisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.

Solubility
Soluble in water, chloroform.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1809-20-7
Molecular Formula C6H14O3P-
Molecular Weight (g/mol) 165.149
MDL Number MFCD00117905
InChI Key IWEDBEZKWHRUJX-UHFFFAOYSA-N
Synonym diisopropylphosphite, acmc-1bsaz, phosphorous acid diisopropyl anion, phosphorous acid, bis 1-methylethyl ester
PubChem CID 21884976
IUPAC Name dipropan-2-yl phosphite
SMILES CC(C)OP([O-])OC(C)C

Specifications

Density 0.99
Boiling Point 72°C to 75°C (10 mmHg)
Flash Point 112°C (234°F)
Linear Formula [(CH3)2CHO]2P(O)H
Refractive Index 1.4075
Quantity 25 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water,chloroform.
Formula Weight 166.16
Percent Purity 98%
Chemical Name or Material Diisopropyl phosphite
Show More Show Less
compliance-icons
Product Identifier
  • Diisopropyl phosphonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Diisopropyl phosphonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diisopropyl phosphite, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.