missing translation for 'onlineSavingsMsg'
Learn More
Diisopropylamine, 99.5%, redistilled, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10308472

Diisopropylamine, 99.5%, redistilled, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10308472
100 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10308472

Brand: Thermo Scientific Acros 378301000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-18-9
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008862
InChI Key UAOMVDZJSHZZME-UHFFFAOYSA-N
Synonym diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
PubChem CID 7912
IUPAC Name N-propan-2-ylpropan-2-amine
SMILES CC(C)NC(C)C

Specifications

Melting Point -61.0°C
Density 0.7160g/mL
Boiling Point 84.0°C
Flash Point −17°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula [(CH3)2CH]2NH
Refractive Index 1.3910 to 1.3930
Quantity 100 mL
Merck Index 15, 3215
Specific Gravity 0.716
Solubility Information Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol,soluble in most common organic solvents
Viscosity 0.4 mPa.s (20°C)
Formula Weight 101.19
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Diisopropylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467563 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog Number378300010,378301000
Lot NumberA0438110 Certificate TypeCertificate of Analysis Date18/10/2023 Catalog Number378300010,378301000
Lot NumberA0444411 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number378300010,378301000
Lot NumberA0356491 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number378300010,378301000
Lot NumberA0413262 Certificate TypeCertificate of Analysis Date18/06/2021 Catalog Number378300010,378301000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diisopropylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diisopropylamine, 99.5%, redistilled, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.