missing translation for 'onlineSavingsMsg'
Learn More
Diisopropylamine, 99%, Thermo Scientific Chemicals Product Code.: 10497160

Diisopropylamine, 99%, Thermo Scientific Chemicals

Product Code. 10497160
1 L
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10497160

Brand: Thermo Scientific Acros 149460010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Diisopropylamine, 99%, C6H15N, CAS Number-108-18-9, 2-propanamine, n-1-methylethyl, bis propan-2-yl amine, diisopropylamine, di-isopropylamine, diisopropyl amine, dipa, n-1-methylethyl-2-propanamine, n-isopropyl-1-amino-2-methylethane, n-isopropylpropan-2-amine, n,n-diisopropylamine, 1L, 04, 154

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-18-9
InChI Key UAOMVDZJSHZZME-UHFFFAOYSA-N
Synonym diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amineShow MoreShow Less
PubChem CID 7912
IUPAC Name N-propan-2-ylpropan-2-amine
SMILES CC(C)NC(C)C

Specifications

Melting Point -61.0°C
Color Colorless
Density 0.7160g/mL
Boiling Point 84.0°C
Flash Point −17°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C6H15N
Linear Formula [(CH3)2CH]2NH
Refractive Index 1.3910 to 1.3930
Quantity 1 L
UN Number 1158
Beilstein 04, 154
Merck Index 15, 3215
Specific Gravity 0.716
Solubility Information Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol, soluble in most common organic solvents
Molecular Weight (g/mol) 101.19
Viscosity 0.4 mPa.s (20°C)
Formula Weight 101.19
Vapor Pressure 95mbar at 20°C
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diisopropylamine, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468285 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number149460010,149460025
Lot NumberA0466517 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number149460010,149460025
Lot NumberA0463783 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number149460010,149460025
Lot NumberA0462476 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog Number149460010,149460025
Lot NumberA0460133 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog Number149460010,149460025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diisopropylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diisopropylamine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.