Learn More
Diisopropylphosphine, 10 wt.% solution in hexanes, Thermo Scientific Chemicals
57.08 € - 591.95 €
Specifications
Chemical Name or Material | Diisopropylphosphine |
---|---|
CAS | 20491-53-6, 110-54-3 |
Color | Brown-Yellow to Yellow |
Density | 0.6700g/mL |
Flash Point | −22°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10539613
|
Thermo Scientific Acros
383400025 |
2.5 g | Glass bottle |
57.08 €
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10756183
|
Thermo Scientific Acros
383400100 |
10 g | Glass bottle |
147.74 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10005752
|
Thermo Scientific Acros
383400500 |
50 g | Glass bottle |
591.95 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diisopropylphosphine | |
Brown-Yellow to Yellow | |
−22°C | |
C6H15P | |
MFCD00233814 | |
diisopropylphosphine, di-isopropylphosphine, di-i-propylphosphine, diisopropylphosphane, phosphine, diisopropyl, phosphine, bis 1-methylethyl, ipr2ph, di-iso-propylphosphine, di-iso-propyl phosphine, di propan-2-yl phosphane | |
WDIIYWASEVHBBT-UHFFFAOYSA-N | |
di(propan-2-yl)phosphane | |
536496 | |
9 to 11% (Total base) |
20491-53-6, 110-54-3 | |
0.6700g/mL | |
Glass bottle | |
[(CH3)2CH]2PH | |
0.67 | |
Solubility in water: insoluble | |
CC(C)PC(C)C | |
118.16 | |
118.16 | |
Liquid |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause damage to organs through prolonged or repeated exposure. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
GHS Signal Word: Danger
- Diisopropylphosphine, 10 wt.% solution in hexanes
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.