Learn More
Dilithium tetrachlorocuprate, 0.1M solution in THF, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 407831000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dilithium tetrachlorocuprate | |
0.1M Solution in THF | |
Cl4CuLi2 | |
MFCD00011081 | |
04,163; 05,226; 06,203; 07,114; 08,176; 11,190; 12,195 | |
HCJWWBBBSCXJMS-UHFFFAOYSA-J | |
dilithium;tetrachlorocopper(2-) | |
11074879 | |
Glass bottle | |
100 mL | |
Liquid |
Colorless | |
15489-27-7, 109-99-9 | |
Li2CuCl4 | |
0.09 to 0.12M | |
0.9100g/mL | |
[Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl | |
219.226 | |
219.24 | |
0.91 | |
−17°C |
Safety and Handling
- Dilithium tetrachlorocuprate, 0.1M solution in THF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only