Learn More
Interscience™ DiluFlow Pro, Single Pump
The DiluFlow Pro automatically dilutes a solid sample in seconds, with the appropriate weight of diluent, up to a total weight of 5 kg. Its robotic arm enables easy work under a laminar flow cabinet.
Brand:  Interscience™ 502105
Product Code. 17895070
  • 8955.00 € / Each
Explore more special offers
Description
Description
  • With the press of only 2 keys, the DiluFlow Pro gravimetric dilutor provides an accurate dilution of all types of samples from 10 to 375 g (at 1/10th)
  • In compliance with ISO 7218 and ISO 6887
  • Robotic arm enables you to place the sample safely in the bag without risk of cross contamination
  • Widest possible opening for the bag and lowest loading threshold
  • With redesigned ergonomics, the ultra-low profile allows optimal comfort of use, especially under a laminar flow hood
  • In case of splashing, the drain tray is removable for rinsing and avoids tedious cleaning
  • GL45 (6.4 mm) per pump
Specifications
Specifications
>99%
Autoclavable
Fast, Standard, Accurate
Diluting Samples
>99%
31.3 x 42.1 x 37.7 cm
60 W
Gravimetric Dilutor
ISO 7218, ISO 6887
100 to 5000 mL
Drip Tray 400, 1 or 2 Pumps, Protection for Distribution Nozzle, Distribution Assembly, USB Cable, Supervision Software, Power Cord, BagOpen 400 for Dilutors, User Manual, Conformity Certificate/Warranty Card, 10 GeckoGrips, 1 Pk. of Adhesive Putty, Allen Wrench, Blender Bag Samples
22.5 mL/sec.
100/240 V
15 A
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Interscience™ DiluFlow Pro, Single Pump > DiluFlow Pro, Single Pump; Type: Gravimetric dilutor