Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for V6 Pumps
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for V6 Pumps has the knowledge and expertise that goes into manufacturing an SGE syringe.
121.00 € - 227.00 €
Specifications
Description | 60mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger |
---|---|
For Use With (Equipment) | V6 Pump |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15218483
|
Trajan Scientific and Medical™
004696 |
50 μL |
150.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15238473
|
Trajan Scientific and Medical™
003696 |
25 μL |
145.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15238493
|
Trajan Scientific and Medical™
006696 |
250 μL |
121.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15238513
|
Trajan Scientific and Medical™
008796 |
5 mL |
137.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15248503
|
Trajan Scientific and Medical™
008196 |
1 mL |
121.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15278513
|
Trajan Scientific and Medical™
008996 |
10 mL |
179.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15288483
|
Trajan Scientific and Medical™
005696 |
100 μL |
121.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15288503
|
Trajan Scientific and Medical™
008596 |
2.5 mL |
133.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15288513
|
Trajan Scientific and Medical™
009496 |
25 mL |
227.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15298493
|
Trajan Scientific and Medical™
007696 |
500 μL |
121.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Diluter/Dispenser Syringe for V6 Pumps
Specifications
60mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger | |
V6 Pump |