missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxydimethylsilane, 97%, Thermo Scientific Chemicals

47.84 € - 130.65 €

Chemical Identifiers

CAS 1112-39-6
Molecular Formula C4H12O2Si
Molecular Weight (g/mol) 120.22
MDL Number MFCD00025691
InChI Key JJQZDUKDJDQPMQ-UHFFFAOYSA-N
Synonym dimethyldimethoxysilane, silane, dimethoxydimethyl, kbm 22, unii-a3qdb1rs1c, dimethyl dimethoxysilane, dimethoxy dimethyl silane, a3qdb1rs1c, dimethoxy-dimethylsilane, dimethyl dimethoxy silane, dimethyl dimethoxy silicane
PubChem CID 66187
IUPAC Name dimethoxydimethylsilane
SMILES CO[Si](C)(C)OC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11489882
View Documents
Thermo Scientific Alfa Aesar
A12025.14
25 g
47.84 €
25g
Please to purchase this item. Need a web account? Register with us today!
11499882
View Documents
Thermo Scientific Alfa Aesar
A12025.22
100 g
130.65 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is employed as silylating agent for diols. Used as a monomer (‘building block’) in the production of silicone polymers or silicone resins, as an intermediate (starting material) in the production of other organosilicon substances. Also employed in non-metal surface treatment to enhance the adhesion properties of surfaces. Used in sealants, in silicone polymer preparations (for the production of elastomers), in mould-making, in semiconductor manufacture, as a laboratory chemical in research and development activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as silylating agent for diols. Used as a monomer (‘building block’) in the production of silicone polymers or silicone resins, as an intermediate (starting material) in the production of other organosilicon substances. Also employed in non-metal surface treatment to enhance the adhesion properties of surfaces. Used in sealants, in silicone polymer preparations (for the production of elastomers), in mould-making, in semiconductor manufacture, as a laboratory chemical in research and development activities.

Solubility
Contact with water releases a flammable alcohol.

Notes
Moisture Sensitive. Store away from oxidizing agents and water/moisture. Protect against electrostatic charges. Keep ignition sources away.
Specifications

Chemical Identifiers

1112-39-6
120.22
JJQZDUKDJDQPMQ-UHFFFAOYSA-N
66187
CO[Si](C)(C)OC
C4H12O2Si
MFCD00025691
dimethyldimethoxysilane, silane, dimethoxydimethyl, kbm 22, unii-a3qdb1rs1c, dimethyl dimethoxysilane, dimethoxy dimethyl silane, a3qdb1rs1c, dimethoxy-dimethylsilane, dimethyl dimethoxy silane, dimethyl dimethoxy silicane
dimethoxydimethylsilane

Specifications

81°C to 82°C
1112-39-6
MFCD00025691
1733694
dimethyldimethoxysilane, silane, dimethoxydimethyl, kbm 22, unii-a3qdb1rs1c, dimethyl dimethoxysilane, dimethoxy dimethyl silane, a3qdb1rs1c, dimethoxy-dimethylsilane, dimethyl dimethoxy silane, dimethyl dimethoxy silicane
JJQZDUKDJDQPMQ-UHFFFAOYSA-N
dimethoxydimethylsilane
66187
97%
Moisture sensitive
Dimethoxydimethylsilane
-80°C
C4H12O2Si
UN1993
−8°C (17°F)
Contact with water releases a flammable alcohol.
CO[Si](C)(C)OC
120.22
120.22
1.371
0.867
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H225-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 214-189-4

missing translation for 'rtecsNumber' : VV3641666

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.