Learn More
Dimethoxymethylsilane, 97%, Thermo Scientific Chemicals
59.60 € - 183.00 €
Chemical Identifiers
CAS | 16881-77-9 |
---|---|
Molecular Formula | C3H9O2Si |
Molecular Weight (g/mol) | 105.188 |
MDL Number | MFCD00048019 |
InChI Key | PKTOVQRKCNPVKY-UHFFFAOYSA-N |
Synonym | methyldimethoxysilane, dimethoxy methyl silane, silane, dimethoxymethyl, dimethoxy methyl silicon, methyl dimethoxysilane, c3h10o2si, methyl dimethoxy silane, dimethoxy methyl silane #, dimethoxy methyl silyl |
PubChem CID | 6329250 |
IUPAC Name | dimethoxy(methyl)silicon |
SMILES | CO[Si](C)OC |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16881-77-9 | |
105.188 | |
PKTOVQRKCNPVKY-UHFFFAOYSA-N | |
6329250 | |
CO[Si](C)OC |
C3H9O2Si | |
MFCD00048019 | |
methyldimethoxysilane, dimethoxy methyl silane, silane, dimethoxymethyl, dimethoxy methyl silicon, methyl dimethoxysilane, c3h10o2si, methyl dimethoxy silane, dimethoxy methyl silane #, dimethoxy methyl silyl | |
dimethoxy(methyl)silicon |
Specifications
61°C | |
16881-77-9 | |
MFCD00048019 | |
2035898 | |
methyldimethoxysilane, dimethoxy methyl silane, silane, dimethoxymethyl, dimethoxy methyl silicon, methyl dimethoxysilane, c3h10o2si, methyl dimethoxy silane, dimethoxy methyl silane #, dimethoxy methyl silyl | |
CO[Si](C)OC | |
105.188 | |
106.2 | |
1.36 | |
0.861 |
-136°C | |
C3H9O2Si | |
UN1993 | |
2°C (35°F) | |
PKTOVQRKCNPVKY-UHFFFAOYSA-N | |
dimethoxy(methyl)silicon | |
6329250 | |
97% | |
Moisture sensitive | |
Dimethoxymethylsilane |
Safety and Handling
GHS H Statement
H225-H318-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 240-914-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only