missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,3-acetonedicarboxylate, 97%, Thermo Scientific Chemicals

Product Code. 10744031
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1830-54-2
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD00008462
InChI Key RNJOKCPFLQMDEC-UHFFFAOYSA-N
Synonym dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
PubChem CID 74591
SMILES COC(=O)CC(=O)CC(=O)OC

Specifications

Melting Point 16.0°C to 17.0°C
Color Colorless to Yellow
Density 1.1800g/mL
Boiling Point 237.0°C to 241.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3O2CCH2COCH2CO2CH3
Refractive Index 1.4430 to 1.4450
Quantity 100 g
Beilstein 03,79
Specific Gravity 1.18
Solubility Information Solubility in water: 120g/L (20°C). Other solubilities: immiscible with ligroin and cyclohexane,soluble in chloroform and ethyl acetate,soluble in acetone,methanol,143g/L 1 n naoh,165g/L 10 % acetic acid
IUPAC Name dimethyl 3-oxopentanedioate
Formula Weight 174.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dimethyl 1, 3-acetonedicarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl 1,3-acetonedicarboxylate
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.