missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 5-bromoisophthalate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 88°C to 90°C |
Quantity | 5 g |
Solubility Information | Insoluble in water. |
Formula Weight | 273.08 |
Percent Purity | 98% |
Chemical Name or Material | Dimethyl 5-bromoisophthalate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'