missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-nitroisophthalate, 98+%, Thermo Scientific Chemicals

Product Code. 11453304
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13290-96-5
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.18
MDL Number MFCD00008429
InChI Key GGTSJKFPGKFLCZ-UHFFFAOYSA-N
Synonym dimethyl 5-nitroisophthalate, 5-nitroisophthalic acid dimethyl ester, 5-nitroisophthalic acid, dimethyl ester, 1,3-benzenedicarboxylic acid, 5-nitro-, dimethyl ester, dimethyl 5-nitro-1,3-benzenedicarboxylate, isophthalic acid, 5-nitro-, dimethyl ester, 1,3-dimethyl 5-nitrobenzene-1,3-dicarboxylate, 5-nitro dimethyl isophthalate, 5-nitro-1,3-benzenedicarboxylic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-nitro-, 1,3-dimethyl ester
PubChem CID 83316
IUPAC Name dimethyl 5-nitrobenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)C(=O)OC)[N+]([O-])=O

Specifications

Melting Point 121°C to 123°C
Quantity 250 g
Beilstein 2140916
Formula Weight 239.19
Percent Purity ≥98%
Chemical Name or Material Dimethyl 5-nitroisophthalate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'