missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl carbonate, 99%, Thermo Scientific Chemicals

Product Code. 10192330
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-38-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00008420
InChI Key IEJIGPNLZYLLBP-UHFFFAOYSA-N
Synonym methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192
PubChem CID 12021
ChEBI CHEBI:36596
IUPAC Name dimethyl carbonate
SMILES COC(=O)OC

Specifications

Melting Point 2.0°C to 4.0°C
Density 1.0690g/mL
Boiling Point 90.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2CO
Refractive Index 1.3672 to 1.3692
Quantity 2.5 kg
Beilstein 03, 4
Fieser 02,149; 05,234; 12,201
Merck Index 15, 3263
Specific Gravity 1.069
Solubility Information Solubility in water: 139g/L. Other solubilities: soluble in all proportions with organic alcohols,soluble in all proportions with esters and ketones,miscible with acids and alkalies
Viscosity 0.625 mPa.s (20°C)
Formula Weight 90.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl carbonate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'