missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl carbonate, 99%, Thermo Scientific Chemicals

Product Code. 11464444
Click to view available options
Quantity:
100 g
500 g
2500 g

Dimethyl carbonate is used as a solvent in organic synthesis and considered as a replacement for solvent like methyl ethyl ketone, tert-butyl acetate and parachlorobenzotrifluoride. It is involved as an intermediate in the preparation of diphenylcarbonate, which in turn is used as a key raw material for the synthesis of Bisphenol-A-polycarbonate. It is also used as a 'green' methylating agent involved in the methylation of aniline, phenols and carboxylic acids. It can be used as a fuel additive due to its high oxygen content. It also finds applications related to supercapacitors and lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl carbonate is used as a solvent in organic synthesis and considered as a replacement for solvent like methyl ethyl ketone, tert-butyl acetate and parachlorobenzotrifluoride. It is involved as an intermediate in the preparation of diphenylcarbonate, which in turn is used as a key raw material for the synthesis of Bisphenol-A-polycarbonate. It is also used as a ′green′ methylating agent involved in the methylation of aniline, phenols and carboxylic acids. It can be used as a fuel additive due to its high oxygen content. It also finds applications related to supercapacitors and lithium batteries.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Air and moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-38-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00008420
InChI Key IEJIGPNLZYLLBP-UHFFFAOYSA-N
Synonym methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192
PubChem CID 12021
ChEBI CHEBI:36596
IUPAC Name dimethyl carbonate
SMILES COC(=O)OC

Specifications

Melting Point ∽4°C
Density 1.070
Boiling Point 90°C
Flash Point 16°C (61°F)
Assay Percent Range 99%
Refractive Index 1.368
Quantity 100 g
UN Number UN1161
Beilstein 635821
Sensitivity Moisture sensitive
Merck Index 14,3241
Solubility Information Miscible with alcohol and ether; Immiscible with water.
Formula Weight 90.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl Carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'