missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl itaconate, 97%, stabilized, Thermo Scientific Chemicals

Product Code. 10061600
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-52-7
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00008446
InChI Key ZWWQRMFIZFPUAA-UHFFFAOYSA-N
Synonym dimethyl itaconate, dimethyl 2-methylenesuccinate, dimethylitaconate, itaconic acid dimethyl ester, itaconic acid, dimethyl ester, butanedioic acid, methylene-, dimethyl ester, 1,4-dimethyl 2-methylidenebutanedioate, dimethyl methylenesuccinate, unii-11jib0yi93, succinic acid, methylene-, dimethyl ester
PubChem CID 69240
SMILES COC(=O)CC(=C)C(=O)OC

Specifications

Melting Point 34.0°C to 39.0°C
Color White
Density 1.2400g/mL
Boiling Point 208.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3O2CCH2C(=CH2)CO2CH3
Quantity 100 g
Beilstein 02,762
Specific Gravity 1.24
Solubility Information Solubility in water: slightly soluble
IUPAC Name dimethyl 2-methylidenebutanedioate
Formula Weight 158.15
Percent Purity 97%
Physical Form Crystalline Low Melting Mass or Liquid
Chemical Name or Material Dimethyl itaconate
Show More Show Less
Product Identifier
  • Dimethyl itaconate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.