Learn More
Dimethyl oxalate, 99%, Thermo Scientific Chemicals
34.40 € - 114.00 €
Chemical Identifiers
CAS | 553-90-2 |
---|---|
Molecular Formula | C4H6O4 |
Molecular Weight (g/mol) | 118.09 |
MDL Number | MFCD00008442 |
InChI Key | LOMVENUNSWAXEN-UHFFFAOYSA-N |
Synonym | methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid |
PubChem CID | 11120 |
ChEBI | CHEBI:6859 |
IUPAC Name | dimethyl oxalate |
SMILES | COC(=O)C(=O)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10347670
|
Thermo Scientific Acros
159271000 |
100 g | Plastic bottle |
34.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10406820
|
Thermo Scientific Acros
159275000 |
500 g | Plastic bottle |
114.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
553-90-2 | |
118.09 | |
LOMVENUNSWAXEN-UHFFFAOYSA-N | |
11120 | |
dimethyl oxalate |
C4H6O4 | |
MFCD00008442 | |
methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid | |
CHEBI:6859 | |
COC(=O)C(=O)OC |
Specifications
553-90-2 | |
Colorless to White | |
163°C | |
Authentic | |
C4H6O4 | |
MFCD00008442 | |
15, 6179 | |
methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid | |
LOMVENUNSWAXEN-UHFFFAOYSA-N | |
dimethyl oxalate | |
11120 | |
118.09 | |
Crystalline Solid |
51°C to 55°C | |
1.1400g/mL | |
75°C | |
98.5% min. (GC) | |
CH3O2CCO2CH3 | |
02, 534 | |
1.14 | |
Solubility in water: 60g/L (25°C). Other solubilities: soluble in ethanol,ether,acetone,chloroform | |
COC(=O)C(=O)OC | |
118.09 | |
CHEBI:6859 | |
99% | |
Dimethyl oxalate, 99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-053-6
missing translation for 'rtecsNumber' : RO2850000
missing translation for 'tsca' : TSCA
RUO – Research Use Only