missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl pimelate, 98+%, Thermo Scientific Chemicals
39.60 € - 356.78 €
Chemical Identifiers
CAS | 1732-08-7 |
---|---|
Molecular Formula | C9H16O4 |
Molecular Weight (g/mol) | 188.22 |
MDL Number | MFCD00008470 |
InChI Key | SHWINQXIGSEZAP-UHFFFAOYSA-N |
Synonym | dimethyl pimelate, heptanedioic acid, dimethyl ester, pimelic acid dimethyl ester, pimelic acid, dimethyl ester, dimethyl 1,7-heptanedioate, 1,7-dimethyl heptanedioate, heptanedioic acid, 1,7-dimethyl ester, unii-ge1pf3049f, heptanedioic acid dimethyl ester, 1ov5vo1 |
PubChem CID | 74416 |
IUPAC Name | dimethyl heptanedioate |
SMILES | COC(=O)CCCCCC(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449507
|
Thermo Scientific Alfa Aesar
B22026.06 |
5 g |
39.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459507
|
Thermo Scientific Alfa Aesar
B22026.14 |
25 g |
118.59 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11469507
|
Thermo Scientific Alfa Aesar
B22026.22 |
100 g |
356.78 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.
Solubility
Soluble in ethanol, benzene, ether. Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Chemical Identifiers
1732-08-7 | |
188.22 | |
SHWINQXIGSEZAP-UHFFFAOYSA-N | |
74416 | |
COC(=O)CCCCCC(=O)OC |
C9H16O4 | |
MFCD00008470 | |
dimethyl pimelate, heptanedioic acid, dimethyl ester, pimelic acid dimethyl ester, pimelic acid, dimethyl ester, dimethyl 1,7-heptanedioate, 1,7-dimethyl heptanedioate, heptanedioic acid, 1,7-dimethyl ester, unii-ge1pf3049f, heptanedioic acid dimethyl ester, 1ov5vo1 | |
dimethyl heptanedioate |
Specifications
1732-08-7 | |
121°C to 122°C (11 mmHg) | |
Mild | |
CH3O2C(CH2)5CO2CH3 | |
MFCD00008470 | |
dimethyl pimelate, heptanedioic acid, dimethyl ester, pimelic acid dimethyl ester, pimelic acid, dimethyl ester, dimethyl 1,7-heptanedioate, 1,7-dimethyl heptanedioate, heptanedioic acid, 1,7-dimethyl ester, unii-ge1pf3049f, heptanedioic acid dimethyl ester, 1ov5vo1 | |
SHWINQXIGSEZAP-UHFFFAOYSA-N | |
dimethyl heptanedioate | |
74416 | |
99% |
1.041 | |
>110°C (230°F) | |
C9H16O4 | |
1.431 | |
1777309 | |
Soluble in ethanol,benzene,ether. Slightly soluble in water. | |
COC(=O)CCCCCC(=O)OC | |
188.22 | |
188.23 | |
Dimethyl pimelate |
Safety and Handling
missing translation for 'einecsNumber' : 217-057-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only