missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl pyrazolo[1,5-a]pyridine-2,3-dicarboxylate, 97%, Thermo Scientific™ Product Code.: 10776915

Dimethyl pyrazolo[1,5-a]pyridine-2,3-dicarboxylate, 97%, Thermo Scientific™

Product Code. 10776915
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10776915

Brand: Thermo Scientific Maybridge MO00792DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5825-71-8
Molecular Formula C11H10N2O4
Molecular Weight (g/mol) 234.211
MDL Number MFCD06411548
InChI Key NZJIUQJPKXRRHB-UHFFFAOYSA-N
Synonym dimethyl pyrazolo 1,5-a pyridine-2,3-dicarboxylate, dimethylpyrazolo 1,5-a pyridine-2,3-dicarboxylate, 2,3-dimethyl pyrazolo 1,5-a pyridine-2,3-dicarboxylate, pyrazolo 1,5-a pyridine-2,3-dicarboxylic acid, dimethyl ester, pyrazolo 1,5-a pyridine-2,3-dicarboxylic acid dimethyl ester, pyrazolo 1,5-a pyridine-2,3-dicarboxylic acid, 2,3-dimethyl ester, pyrazolo 1,5-a pyridine-2,3-dicarboxylicacid, 2,3-dimethyl ester, methyl 3-methoxycarbonyl-8-hydropyrazolo 1,5-a pyridine-2-carboxylateShow MoreShow Less
PubChem CID 2794621
IUPAC Name dimethyl pyrazolo[1,5-a]pyridine-2,3-dicarboxylate
SMILES COC(=O)C1=C2C=CC=CN2N=C1C(=O)OC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 234.21
Percent Purity 97%
Chemical Name or Material Dimethyl pyrazolo[1, 5-a]pyridine-2, 3-dicarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl pyrazolo[1,5-a]pyridine-2,3-dicarboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.