missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfOxide, ≥99.9% (GC), Honeywell Riedel-de Haën™

Product Code. 15615740
Click to view available options
Quantity:
100 mL
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

CHROMASOLV™, GC-Headspace tested,≥99.9%

Honeywell Chromasolv™ GC-headspace solvents:

  • developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • very pure solvents with extremely low concentrations of the defined residual solvents
  • state-of-the art production and application tests

 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Density 1.10g/mL
Quantity 100 mL
Explosion Limit 42%, 63°F
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Linear Formula (CH3)2SO
Vapor Pressure 0.42 mmHg (20°C)
UN Number NA1993
IUPAC Name methanesulfinylmethane
Formula Weight 78.13
Percent Purity ≥99.9%
Grade CHROMASOLV™
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'