missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals Product Code.: 11495170

Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals

Product Code. 11495170
500 mL
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 x 4 L
This item is not returnable. View return policy

Product Code. 11495170

Brand: Thermo Scientific Alfa Aesar 036480.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. Used in antifreeze or hydraulic fluids. Utilized in the oxidation of thiols and disulfides to sulfonic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
Density 1.101 g/mL
Boiling Point 189°C
Quantity 500 mL
Flash Point 87°C (189°F)
Physical Form Clear,colorless Liquid
Refractive Index 1.479
Assay Percent Range 99.9%
Beilstein 506008
Solubility Information Miscible with water,diethyl ether,ethanol,acetone,ether,carbon tetrachloride and ethyl acetate.
Formula Weight 78.13
Grade ACS
Chemical Name or Material Dimethyl sulfoxide, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl sulfoxide, ACS, 99.9% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.