missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl Sulfoxide, for HPLC, Fisher Chemical™ Product Code.: 10122140

Dimethyl Sulfoxide, for HPLC, Fisher Chemical™

Product Code. 10122140
500 mL
Click to view available options
Quantity:
250 mL
500 mL
2.5 L
This item is not returnable. View return policy

Product Code. 10122140

Brand: Fisher Chemical D/4125/PB08

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
Boiling Point 189°C
Quantity 500 mL
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 0.55mbar at 20°C
Viscosity 1.98 mPaS at 25°C
IUPAC Name methylsulfinylmethane
Formula Weight 78.13g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2527008 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberD/4125/PB08,D/4125/PB07,D/4125/PB17,D/4125/99
Product Identifier
  • Methyl sulfoxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl Sulfoxide, for HPLC, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.