missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide Molecular Biology MP Biomedicals

Product Code. 11456521
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 67-68-5
Density 1.104 g/cm3 at 20 °C
Quantity 100 mL
Molecular Formula C2H6OS
MDL Number MFCD00002089
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES CS(C)=O
IUPAC Name methylsulfinylmethane
Molecular Weight (g/mol) 78.13
PubChem CID 679
ChEBI CHEBI:28262
Formula Weight 78.13
Grade Molecular Biology
Chemical Name or Material Dimethyl Sulfoxide
Show More Show Less
Product Identifier
  • Dimethyl Sulfoxide, molecular biology reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.