missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl trans-1,2-cyclopropanedicarboxylate, 99+%

Product Code. 10328860 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10328860

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 826-35-7 | C7H10O4 | 158.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 826-35-7
Color Colorless
Density 1.1200g/mL
Boiling Point 101°C (18.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C7H10O4
Refractive Index 1.4425 to 1.4445
MDL Number MFCD00062806
Quantity 5 g
Specific Gravity 1.12
Synonym dimethyl trans-1,2-cyclopropanedicarboxylate, 1r,2r-rel-dimethyl cyclopropane-1,2-dicarboxylate, dimethyl 1r,2r-cyclopropane-1,2-dicarboxylate, cyclopropane-1,2-dicarboxylic acid dimethyl ester, e, dimethyltrans-1,2-cyclopropanedicarboxylate, dimethyl-trans-1,2-cyclopropanedicarboxylate, 1,2-dimethyl 1r,2r-cyclopropane-1,2-dicarboxylate, dimethyl +/--trans-cyclopropane-1,2-dicarboxylate, trans-cyclopropane-1,2-dicarboxylic acid dimethylester, 1r,2r ;-rel-dimethyl cyclopropane-1,2-dicarboxylate
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in all organic solvents
InChI Key JBVOSZYUSFDYIN-RFZPGFLSSA-N
SMILES COC(=O)C1CC1C(=O)OC
IUPAC Name dimethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
Molecular Weight (g/mol) 158.16
PubChem CID 6429599
Formula Weight 158.16
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dimethyl trans-1, 2-cyclopropanedicarboxylate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl trans-1,2-cyclopropanedicarboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.