missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl trisulfide, 98+%

Product Code. 10289500 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10289500

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3658-80-8 | C2H6S3 | 126.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3658-80-8
Molecular Formula C2H6S3
Molecular Weight (g/mol) 126.27
MDL Number MFCD00039808
InChI Key YWHLKYXPLRWGSE-UHFFFAOYSA-N
Synonym dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts
PubChem CID 19310
ChEBI CHEBI:4614
IUPAC Name (methyltrisulfanyl)methane
SMILES CSSSC

Specifications

Melting Point -68.0°C
Color Yellow
Density 1.1900g/mL
Boiling Point 165.0°C to 175.0°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3SSSCH3
Refractive Index 1.5995 to 1.6025
Quantity 25 g
Beilstein 01, IV, 1283
Specific Gravity 1.19
Formula Weight 126.27
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Dimethyl trisulfide, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethyl trisulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.