missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals
Click to view available options
Quantity:
10 Ea.
50 Ea.
Description
Dimethylamine is a used as a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber. It is raw material for the production of many agrochemicals and pharmaceuticals, such as dimefox and diphenhydramine, respectively.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Density | 0.89 |
Flash Point | 15°C (60°F) |
Packaging | Ampoule |
Refractive Index | 1.37 |
Quantity | 10 Ea. |
UN Number | UN1160 |
Beilstein | 605257 |
Specific Gravity | 0.89 |
Solubility Information | Fully miscible in water. |
Formula Weight | 45.08 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'