Learn More
Dimethylamine, 40 wt.% solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 163670025
54.55 EUR valid until 2024-12-31
Use promo code "21665" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethylamine | |
-37.0°C | |
0.8900g/mL | |
−18°C | |
C2H7N | |
1.37 | |
2.5 L | |
07,119 | |
0.89 | |
Solubility in water: >500g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CNC | |
45.07 | |
CHEBI:17170 | |
≥40% |
124-40-3, 7732-18-5 | |
Colorless | |
54.0°C | |
Glass bottle | |
(CH3)2NH | |
MFCD00008288 | |
04, 39 | |
15, 3250 | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
N-methylmethanamine | |
674 | |
45.07 | |
Liquid |
Safety and Handling
- Dimethylamine, solution in water
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- Flammable liquid Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Dimethylamine
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only