Learn More
Dimethylaminoacetonitrile, 98%, Thermo Scientific Chemicals
34.60 € - 86.10 €
Chemical Identifiers
CAS | 926-64-7 |
---|---|
Molecular Formula | C4H8N2 |
Molecular Weight (g/mol) | 84.12 |
MDL Number | MFCD00001890 |
InChI Key | PLXBWEPPAAQASG-UHFFFAOYSA-N |
Synonym | dimethylaminoacetonitrile, dimethylamino acetonitrile, acetonitrile, dimethylamino, 2-dimethylamino acetonitrile, n-cyanomethyl dimethylamine, glycinonitrile, n,n-dimethyl, unii-wu34f15n7c, glycinonitrile,n-dimethyl |
PubChem CID | 61237 |
IUPAC Name | 2-(dimethylamino)acetonitrile |
SMILES | CN(C)CC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309889
|
Thermo Scientific Alfa Aesar
B25085.14 |
25 g |
34.60 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319889
|
Thermo Scientific Alfa Aesar
B25085.22 |
100 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
926-64-7 | |
84.12 | |
PLXBWEPPAAQASG-UHFFFAOYSA-N | |
61237 | |
CN(C)CC#N |
C4H8N2 | |
MFCD00001890 | |
dimethylaminoacetonitrile, dimethylamino acetonitrile, acetonitrile, dimethylamino, 2-dimethylamino acetonitrile, n-cyanomethyl dimethylamine, glycinonitrile, n,n-dimethyl, unii-wu34f15n7c, glycinonitrile,n-dimethyl | |
2-(dimethylamino)acetonitrile |
Specifications
926-64-7 | |
36°C (96°F) | |
C4H8N2 | |
MFCD00001890 | |
1735677 | |
PLXBWEPPAAQASG-UHFFFAOYSA-N | |
2-(dimethylamino)acetonitrile | |
61237 | |
98% |
137°C to 138°C | |
Recognizable | |
(CH3)2NCH2CN | |
UN2378 | |
dimethylaminoacetonitrile, dimethylamino acetonitrile, acetonitrile, dimethylamino, 2-dimethylamino acetonitrile, n-cyanomethyl dimethylamine, glycinonitrile, n,n-dimethyl, unii-wu34f15n7c, glycinonitrile,n-dimethyl | |
CN(C)CC#N | |
84.12 | |
84.12 | |
Dimethylaminoacetonitrile |
Safety and Handling
GHS H Statement
H300-H310-H226-H319
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapour.
Causes serious eye irritation.
GHS P Statement
P260-P280h-P305+P351+P338-P309-P310-P302+P352
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-DIMETHYLAMINOACETONITRILE
missing translation for 'rtecsNumber' : AL9450000
RUO – Research Use Only