missing translation for 'onlineSavingsMsg'
Learn More

Dimethylethoxysilane, 94%, Thermo Scientific Chemicals

Product Code. 10339353
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14857-34-2
Molecular Formula C4H12OSi
Molecular Weight (g/mol) 104.22
MDL Number MFCD00026750
InChI Key DRUOQOFQRYFQGB-UHFFFAOYSA-N
Synonym ethoxydimethylsilane, dimethylethoxysilane, silane, ethoxydimethyl, dimethyl ethoxysilane, ethoxy dimethyl silicon, dimethylethoxysilane, ammonia-free, ethoxydimethylsilyl, ethoxydimethyl-silan, sgqhhgcihcup@, 4-04-00-03991 beilstein handbook reference
PubChem CID 6328550
IUPAC Name ethoxy(dimethyl)silicon
SMILES CCO[Si](C)C

Specifications

Color Colorless
Boiling Point 54.0°C
Flash Point −5°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2OSiH(CH3)2
Refractive Index 1.3670 to 1.3690 (20°C, 589nm)
Quantity 5 g
Solubility Information Solubility in water: reacts
Formula Weight 104.22
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Dimethylethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethylethoxysilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'