missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime, 99+%

Product Code. 10012170 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10012170

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dimethylglyoxime, 99+%, C4H8N2O2, CAS Number-95-45-4, 2,3-butanedione dioxime, chugaev's reagent, biacetyl, dioxime, 2,3-diisonitrosobutane, diacetyldioxime, nsc 9, diacetyl dioxime, unii-2971mft1ky, biacetyl dioxime, dimethylglyoxime, 500g, 01,772, White, 202-420-1, 116.12, GHS Signal Word: Danger | CAS: 95-45-4 | C4H8N2O2 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-45-4
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002117
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
PubChem CID 5356010
SMILES C\C(=N/O)\C(\C)=N\O

Specifications

Melting Point 239.0°C to 242.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula CH3C(=NOH)C(=NOH)CH3
Quantity 500 g
Beilstein 01,772
Merck Index 15,327
Solubility Information Solubility in water: 0,6g/L 20°C. Other solubilities: soluble in alcohol,ether,pyridine and acetone,slightly soluble in benzene
Sublimation Point >70 °C
IUPAC Name N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine
Formula Weight 116.12
Percent Purity 99+%
Insoluble Matter 0.05% max. (in alcohol)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylglyoxime, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.