missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylglyoxime, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Dimethylglyoxime is a well known reagent in analytical chemistry. It is used for the qualitative and quantitative determination of nickel by gravimetric analysis. It also finds application in the separation of palladium from tin, gold, rhodium and iridium. It is also useful in the detection of bismuth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethylglyoxime is a well known reagent in analytical chemistry. It is used for the qualitative and quantitative determination of nickel by gravimetric analysis. It also finds application in the separation of palladium from tin, gold, rhodium and iridium. It is also useful in the detection of bismuth.
Solubility
Soluble in alcohol, ether and solutions of alkalies. Insoluble in water.
Notes
Incompatible with oxidizing agents.
Dimethylglyoxime is a well known reagent in analytical chemistry. It is used for the qualitative and quantitative determination of nickel by gravimetric analysis. It also finds application in the separation of palladium from tin, gold, rhodium and iridium. It is also useful in the detection of bismuth.
Solubility
Soluble in alcohol, ether and solutions of alkalies. Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | ∼240°C (decomposition) |
Quantity | 100 g |
Beilstein | 506731 |
Merck Index | 14,3246 |
Solubility Information | Soluble in alcohol,ether and solutions of alkalies. Insoluble in water. |
Formula Weight | 116.12 |
Percent Purity | 99% |
Chemical Name or Material | Dimethylglyoxime |
Safety and Handling
Product Identifier
- Dimethylglyoxime
- Danger
- Acute toxicity Category 3
- Flammable solid Category 2
- H228-Flammable solid.
- H301-Toxic if swallowed.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P405-Store locked up.
- MIXTURE LIST-Contains : 2,3-Butanedione, dioxime
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'