missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylphenylsilane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Beschreibung
Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.
Solubility
Not miscible in water.
Notes
Moisture sensitive. Incompatible materials are Strong oxidizing agents, Strong acids.
Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.
Solubility
Not miscible in water.
Notes
Moisture sensitive. Incompatible materials are Strong oxidizing agents, Strong acids.
Spezifikation
Spezifikation
Boiling Point | 156°C to 157°C |
Melting Point | -124°C |
Quantity | 10 g |
UN Number | UN1993 |
Beilstein | 2204906 |
Flash Point | 35°C (95°F) |
Solubility Information | Not miscible in water. |
Formula Weight | 136.27 |
Percent Purity | 97% |
Refractive Index | 1.497 |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'