missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylphenylsilanol sodium salt, 97%, Thermo Scientific Chemicals
84.70 € - 250.00 €
Chemical Identifiers
CAS | 7646-75-5 |
---|---|
Molecular Formula | C8H11NaOSi |
Molecular Weight (g/mol) | 174.25 |
MDL Number | MFCD09266151 |
InChI Key | NPZJKOMUCXNLBN-UHFFFAOYSA-N |
Synonym | dimethylphenylsilanol sodium salt, sodium dimethyl phenyl silanolate, sodium dimethylphenylsilanolate, amtsi046, sodium phenyldimethylsilanolate, dimethyl phenyl sodiooxy silane, sodium phenyl dimethylsilanolate |
PubChem CID | 23713317 |
IUPAC Name | sodium;dimethyl-oxido-phenylsilane |
SMILES | C[Si](C)(C1=CC=CC=C1)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11462050
|
Thermo Scientific Alfa Aesar
H28457.03 |
1 g |
84.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11472050
|
Thermo Scientific Alfa Aesar
H28457.06 |
5 g |
250.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7646-75-5 | |
174.25 | |
NPZJKOMUCXNLBN-UHFFFAOYSA-N | |
23713317 | |
C[Si](C)(C1=CC=CC=C1)[O-].[Na+] |
C8H11NaOSi | |
MFCD09266151 | |
dimethylphenylsilanol sodium salt, sodium dimethyl phenyl silanolate, sodium dimethylphenylsilanolate, amtsi046, sodium phenyldimethylsilanolate, dimethyl phenyl sodiooxy silane, sodium phenyl dimethylsilanolate | |
sodium;dimethyl-oxido-phenylsilane |
Specifications
7646-75-5 | |
MFCD09266151 | |
NPZJKOMUCXNLBN-UHFFFAOYSA-N | |
sodium;dimethyl-oxido-phenylsilane | |
23713317 | |
97% |
C8H11NaOSi | |
dimethylphenylsilanol sodium salt, sodium dimethyl phenyl silanolate, sodium dimethylphenylsilanolate, amtsi046, sodium phenyldimethylsilanolate, dimethyl phenyl sodiooxy silane, sodium phenyl dimethylsilanolate | |
C[Si](C)(C1=CC=CC=C1)[O-].[Na+] | |
174.25 | |
174.25 | |
Dimethylphenylsilanol sodium salt |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only