missing translation for 'onlineSavingsMsg'
Learn More

Dimethyltin dichloride, 98%, Thermo Scientific Chemicals

71.20 € - 227.00 €

Chemical Identifiers

CAS 753-73-1
Molecular Formula C2H6Cl2Sn
Molecular Weight (g/mol) 219.68
MDL Number MFCD00000517
InChI Key PKKGKUDPKRTKLJ-UHFFFAOYSA-L
Synonym dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
PubChem CID 12955
IUPAC Name dichloro(dimethyl)stannane
SMILES C[Sn](C)(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11443238
View Documents
Thermo Scientific Alfa Aesar
A17661.06
5 g
71.20 €
5g
Please to purchase this item. Need a web account? Register with us today!
11453238
View Documents
Thermo Scientific Alfa Aesar
A17661.14
25 g
227.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

753-73-1
219.68
PKKGKUDPKRTKLJ-UHFFFAOYSA-L
12955
C[Sn](C)(Cl)Cl
C2H6Cl2Sn
MFCD00000517
dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
dichloro(dimethyl)stannane

Specifications

753-73-1
188°C to 190°C
MFCD00000517
3660508
dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
C[Sn](C)(Cl)Cl
219.68
219.69
Dimethyltin dichloride
104°C to 108°C
C2H6Cl2Sn
UN3146
Moisture sensitive
PKKGKUDPKRTKLJ-UHFFFAOYSA-L
dichloro(dimethyl)stannane
12955
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311-H314-H330-H360-H372

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 212-039-2

missing translation for 'rtecsNumber' : WH7245000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only