missing translation for 'onlineSavingsMsg'
Learn More

Dimethyltin dichloride, Thermo Scientific Chemicals

Product Code. 11344829
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a pharmaceutical intermediate. It is employed as a coating, plastic and rubber auxilliary agent. Used as methyl Tin stabilizer intermediates, catalysts for esterification reaction, raw material for glass coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 753-73-1
Molecular Formula C2H6Cl2Sn
Molecular Weight (g/mol) 219.68
MDL Number MFCD00000517
InChI Key PKKGKUDPKRTKLJ-UHFFFAOYSA-L
Synonym dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
PubChem CID 12955
IUPAC Name dichloro(dimethyl)stannane
SMILES C[Sn](C)(Cl)Cl

Specifications

Melting Point 103°C to 105°C
Boiling Point 185°C to 190°C
Quantity 10 g
UN Number UN3146
Beilstein 3660508
Sensitivity Moisture Sensitive
Formula Weight 219.69
Physical Form Crystalline
Chemical Name or Material Dimethyltin dichloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'