Learn More
OHAUS™ Dimpled Mat
OHAUS™ Dimpled Mat designed to securely hold tubes in place.
Brand:  OHAUS™ 30400144
 View more versions of this product
Product Code. 15762559
  • 56.40 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
  • Platforms can increase the amount of usable space for samples
  • In some cases can be stacked to increase throughput and double sample capacity
  • Tube racks and dimpled mats securely hold tubes in place
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Platform Mat
14.00 in.
35.6 x 28 cm
35.60 cm
Dimpled Mat For Rocker SHRK07AL
PVC
35.60 cm
14.00 in.
28.00 cm
Rocker