Learn More
Lenz Laborglasintrumente Dimroth Condensers for Extractors with GL Threads
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
81.00 € - 198.00 €
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
10101602
|
Lenz Laborglasintrumente
05246015 |
NS 34/35 |
95.00 €
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||||
10530583
|
Lenz Laborglasintrumente
05246014 |
NS 29/32 |
81.00 €
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||||
10355042
|
Lenz Laborglasintrumente
05246018 |
NS 71/51 |
198.00 €
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||||
10782903
|
Lenz Laborglasintrumente
05246017 |
NS 60/46 |
159.00 €
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||||
Description
- Dimroth condensers for extractors
- With GL threads Dimroth condensers for extractors
- For condensing vapours
- With NS cone
- Water connection: GL 14 thread (with two threaded plastic olives)
- Made of DURAN™ tubing
Specifications
Duran Glass |