Learn More
Lenz Laborglasintrumente Dimroth Condensers, with Glass Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05240201
 View more versions of this product
Product Code. 10047770
  • 106.00 € / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Dimroth condensers acc. to DIN 12591
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Specifications
160 mm
NS 14/23
Duran Glass