missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dioctyl sulfosuccinate, sodium salt, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Plastic Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Acidity | 2.5 max. (on solids basis) |
Melting Point | 153.0°C to 157.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min |
Packaging | Plastic Bottle |
Quantity | 1 kg |
Beilstein | 04, IV, 114 |
Fieser | 15,149 |
Merck Index | 15, 3446 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0471725 | Certificate of Analysis | 11/04/2025 | 117100010,117101000,117100025 |
A0467087 | Certificate of Analysis | 31/10/2024 | 11710035,117100010,117101000,117100025 |
A0425907 | Certificate of Analysis | 05/04/2024 | 11710035,117100010,117101000,117100025 |
A0447844 | Certificate of Analysis | 25/01/2024 | 11710035,117100010,117101000,117100025 |
A0452279 | Certificate of Analysis | 25/01/2024 | 11710035,117100010,117101000,117100025 |
Safety and Handling

Product Identifier
- Dioctyl sulfosuccinate, sodium salt
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dioctyl sulfosuccinate, sodium salt, 96%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction