Learn More
Thermo Scientific™ Dionex™ AS-AP Autosampler Vial Kits
Use Thermo Scientific™ Dionex™ AS-AP Autosampler Vial Kits offered in polystyrene, glass or polypropylene, with caps and split septa.
Brand:  Thermo Scientific™ 055058
 View more versions of this product
Product Code. 11371743
  • 185.00 € / Pack of 100
Explore more special offers
Description
Description
Use Thermo Scientific™ Dionex™ AS-AP Autosampler Vial Kits offered in polystyrene, glass or polypropylene, with caps and split septa.
Septa included are blue.
Specifications
Specifications
Vial Kit
Polystyrene
10 mL
Caps and Split Septa
AS-AP Autosampler
Each 1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Dionex™ AS-AP Autosampler Vial Kits > Polystyrene; 10mL; With caps and septa
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.