Learn More
Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns

Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
Brand: Thermo Scientific™ 063000
1228.70 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
- Achieve n, n-1 resolution for oligonucleotides
- Resolve oligonucleotides with secondary structures
- Assay phosphorothioate purity
- Selectivity control with eluent pH, salt, and solvent
- Resolve RNA with aberrant (2' – 5') links from normal SSRNA

Specifications
Analytical Column | |
1.2 mL/min | |
250 mm | |
pH 2 to 12.5, 100% compatible with common organic solvents, Ionic form eluents: chloride, perchlorate | |
2.5 to 12.5 | |
Each | |
Quaternary Ammonium | |
Spherical, Nonporous Polymer | |
Ion Exchange | |
Bio-LC |
4 mm | |
UltiMate 3000 Titanium System | |
4000 psi (275 bar) | |
8 μm | |
Nonporous | |
85°C | |
DNAPac | |
Ion Exchange | |
Analytical Column | |
Responsibly packaged |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.