missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns

Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.

Brand:  Thermo Scientific™ 082510

2799.55 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15139673

  • 2813.55 € / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.

  • Achieve n, n-1 resolution for oligonucleotides
  • Resolve oligonucleotides with secondary structures
  • Assay phosphorothioate purity
  • Selectivity control with eluent pH, salt, and solvent
  • Resolve RNA with aberrant (2' – 5') links from normal SSRNA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Analytical Column
UltiMate 3000 Titanium System
10000 psi (690 bar)
2.5 to 12.5
Each
Quaternary Ammonium
Spherical, Nonporous Polymer
Ion Exchange
Bio-LC
4.6 mm
250 mm
4 μm
Nonporous
85°C
DNAPac
Ion Exchange
Analytical Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.