missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dionex™ IonPac™ NS1 Analytical & Guard Columns

Achieve high-efficiency separations of hydrophobic, ionizable compounds with these polymeric-based, reversed-phase columns.

Brand:  Thermo Scientific™ 088763

 View more versions of this product

Product Code. 15530224

  • 272.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Achieve high-efficiency separations of hydrophobic, ionizable compounds with Thermo Scientific™ Dionex™ IonPac™ NS1 10μm Analytical & Guard Columns. The columns of choice for ion-pair or ion-suppression chromatography, these polymer-based, reversed-phase columns provide excellent resolution and peak shape. Packing is a neutral, macroporous, high surface area, ethylvinylbenzene polymer cross linked with 55% divinylbenzene, making IonPac NS1 columns resistant to solvents, acids and bases while permitting the use of eluent from pH 0 to 14.

Polymer-Based Columns for Efficient Reversed-Phase Separations
  • Ideal for separation of large molecules that carry localized charges, such as surfactants
  • Can be used for traditional polymeric reversed-phase applications
  • Use with ion-pair chromatography for difficult separations
  • Available in 5 and 10μm particle sizes
Key Applications

Use IonPac NS1 columns for reliable separations of various compounds: High molecular weight aliphatic, carboxylic acids; Alkyl and aromatic sulfates and sulfonates; Quaternary ammonium ions; Water-soluble vitamins; Sulfur oxides; Metal cyanide complexes; Phenols

Specifications

Specifications

Mixed Mode
Reversed Phase, Ion-Pairing, Ion-Suppression
10 μm
Divinylbenzene
0 to 14
2 mm
1 mL/min
300 psi (20 bar)
ProPac
Macroporous Ethylvinylbenzene Polymer
50 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.