Learn More
Thermo Scientific™ Dionex™ MAbPac™SCX-10 Ion Exchange Columns
For the high-resolution separation and analysis of monoclonal antibody variants
3990.00 € - 4009.52 €
Specifications
Type Bio-LC
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
15530244
View Documents
Thermo Scientific™
088783
3 Lots of Resin
4009.52 €
Each
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
15520244
View Documents
Thermo Scientific™
088782
1 Lot of Resin This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Unequalled high-resolution separations
  • Exceptionally high efficiency
  • Highest resolution of monoclonal antibody variants
  • Unmatched column-to-column and lot-to-lot reproducibility
  • Hydrophobic interactions eliminated
  • Excellent for stability testing
Specifications
Specifications
Bio-LC