missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ OmniPac™ PAX-500 HPLC Columns

When you need to separate anionic and neutral species in a single run, use this solvent-compatible mixed-mode column.

593.74 € - 1577.63 €

Specifications

Column Type Mixed Mode
For Use With (Application) Reversed Phase and Ion Exchange
Max. Pressure 4000 psi (275 bar)
Particle Size 8.5 μm
Stationary Phase Alkanol Quaternary Amine
View More Specs

Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
11301723
View Documents
Thermo Scientific™
042152
250 mm
1577.63 €
Each
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
 
11311723
View Documents
Thermo Scientific™
042153
50 mm
593.74 €
Each
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
 
Description

Description

When you need to separate anionic and neutral species in a single run, use the Thermo Scientific™ OmniPac™ PAX-500 Column. With anion-exchange and reversed-phase properties, this polymer-based mixed-mode column is solvent compatible and stable over the entire pH range of 1-14. Solvents may be used to affect selectivity as well as to remove contaminants to extend column lifetime. The OmniPac PAX-500 column is ideal for analysis of a wide range of compounds.

The OmniPac PAX-500 column is ideal for analysis of alcohols, alkanolamines, antihistamines, anti-inflammatory agents, aromatic acids, inorganic acids, peptides, purines and pyrimidines, sulfonamides, sulfonated anionic surfactants, and water-soluble vitamins.
  • Macroporous hydrophobic core provides reversed-phase retention and is coated with an anion-exchange latex bead layer for anion-exchange retention
  • Ion exchange capacity of approximately 40μeq/column
  • 100% solvent-compatible and should always be run with at least 1% organic solvent in the mobile phase
Specifications

Specifications

Mixed Mode
4000 psi (275 bar)
Alkanol Quaternary Amine
0 to 14
Each
L50
Reversed Phase and Ion Exchange
8.5 μm
Spherical, Microporous Polymer
1200 Å
300 m²/g
4 mm
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.