Learn More
Thermo Scientific™ Dionex™ PEEK Tubing Cutter
The Thermo Scientific™ Dionex PEEK Tubing Cutter has a guillotine with 1/16 in. and 1/8 in. guide holes.
Brand:  Thermo Scientific™ 160484
Product Code. 11312133
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The Thermo Scientific™ Dionex PEEK Tubing Cutter has a guillotine with 1/16 in. and 1/8 in. guide holes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PEEK tubing cutter
Tubing
nanoLC
Tubing Cutter