Learn More
Thermo Scientific™ Dionex™ Specially Formulated PTFE Tubing
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC). Choose fitting dimensions for OD of fused silica tubing used.
Brand:  Thermo Scientific™ 160489
 View more versions of this product
Product Code. 11352133
  • 48.05 € / Pack of 2
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 and 350 um ID
Fused Silica Capillary Tubing
nanoLC
Capillary Tubing