missing translation for 'onlineSavingsMsg'
Learn More

Dioxane solution EU Pharmacopoeia, Fisher chemical™
BIOPHARMA

Product Code. 12940404 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Packaging:
Bottle
Purity Grade Notes:
Eu Ph Ref: 1032001
Eu Ph Ref: 1032002
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 12940404

Brand: Fisher Chemical RP 066 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
Type Mass Spectrometry Standards
Quantity 100 mL
Grade Eu Ph Ref: 1032001
Chemical Name or Material Dioxan stock solution.
compliance-icons
Product Identifier
  • Dioxan stock solution. European Pharmacopoeia Ref: 1032001
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,4-dioxane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.