missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl-2-pyridylphosphine, 98%, Thermo Scientific Chemicals

108.00 € - 378.00 €

Chemical Identifiers

CAS 37943-90-1
Molecular Formula C17H14NP
Molecular Weight (g/mol) 263.28
MDL Number MFCD00192108
InChI Key SVABQOITNJTVNJ-UHFFFAOYSA-N
Synonym diphenyl-2-pyridylphosphine, 2-diphenylphosphino pyridine, 2-pyridyldiphenylphosphine, diphenyl 2-pyridinyl phosphine, diphenyl 2-pyridyl phosphane, 2-diphenylphosphanyl-pyridine, 2-diphenylphosphanyl pyridine, diphenyl pyridin-2-yl phosphane, pph2 2-pyridyl, dpppy
PubChem CID 621893
IUPAC Name diphenyl(pyridin-2-yl)phosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=N3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15412458
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H56029.06
5 g
108.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
15422458
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H56029.14
25 g
378.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.

Solubility
Soluble in toluene. (almost transparency)

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Chemical Identifiers

37943-90-1
263.28
SVABQOITNJTVNJ-UHFFFAOYSA-N
621893
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=N3
C17H14NP
MFCD00192108
diphenyl-2-pyridylphosphine, 2-diphenylphosphino pyridine, 2-pyridyldiphenylphosphine, diphenyl 2-pyridinyl phosphine, diphenyl 2-pyridyl phosphane, 2-diphenylphosphanyl-pyridine, 2-diphenylphosphanyl pyridine, diphenyl pyridin-2-yl phosphane, pph2 2-pyridyl, dpppy
diphenyl(pyridin-2-yl)phosphane

Specifications

37943-90-1
C17H14NP
diphenyl-2-pyridylphosphine, 2-diphenylphosphino pyridine, 2-pyridyldiphenylphosphine, diphenyl 2-pyridinyl phosphine, diphenyl 2-pyridyl phosphane, 2-diphenylphosphanyl-pyridine, 2-diphenylphosphanyl pyridine, diphenyl pyridin-2-yl phosphane, pph2 2-pyridyl, dpppy
SVABQOITNJTVNJ-UHFFFAOYSA-N
diphenyl(pyridin-2-yl)phosphane
621893
98%
82°C to 84°C
MFCD00192108
Soluble in toluene. (almost transparency)
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=N3
263.28
263.27
Diphenyl-2-pyridylphosphine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only